Assay Details
| Gene Symbol: SWP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111346.4 | NP_187125.1 | 6 - 7 | 3522 | 65 | |
| GenBank mRNA | AF466359.1 | - | 6 - 7 | 3584 | 65 | |
| AJ511271.1 | - | 6 - 7 | 3584 | 65 | ||
More Information
Set Membership: |
|

SWP
Genomic Map