Assay Details
| Gene Symbol: ZDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337544.1 | NP_001319473.1 | 12 - 13 | 1640 | 81 | |
| NM_001337545.1 | NP_001319474.1 | 12 - 13 | 1518 | 81 | ||
| NM_111359.4 | NP_187138.1 | 11 - 12 | 1609 | 81 | ||
| NM_202493.3 | NP_974222.1 | 12 - 13 | 1505 | 81 | ||
| GenBank mRNA | AY059920.1 | - | 11 - 12 | 1470 | 81 | |
| AY072096.1 | - | 11 - 12 | 1476 | 81 | ||
| AY096583.1 | - | 11 - 12 | 1362 | 81 | ||
More Information
Set Membership: |
|

ZDS
Genomic Map