Assay Details
| Gene Symbol: NLP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341638.1 | NP_001328405.1 | 3 - 4 | 1147 | 68 | |
| NM_118534.6 | NP_194133.1 | 2 - 3 | 1281 | 68 | ||
More Information
Set Membership: |
|

NLP7
Genomic Map