Assay Details
| Gene Symbol: AGL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118587.6 | NP_194185.1 | 2 - 3 | 405 | 113 | |
| GenBank mRNA | AB493697.1 | - | 1 - 2 | 176 | 113 | |
| AF005158.2 | - | 2 - 3 | 242 | 113 | ||
| BT025171.1 | - | 1 - 2 | 176 | 113 | ||
More Information
Set Membership: |
|

AGL24
Genomic Map