Assay Details
| Gene Symbol: AGL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118587.6 | NP_194185.1 | 3 - 4 | 495 | 124 | |
| GenBank mRNA | AB493697.1 | - | 2 - 3 | 266 | 124 | |
| AF005158.2 | - | 3 - 4 | 332 | 124 | ||
| BT025171.1 | - | 2 - 3 | 266 | 124 | ||
More Information
Set Membership: |
|

AGL24
Genomic Map