Assay Details
| Gene Symbol: AGL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118587.6 | NP_194185.1 | 5 - 6 | 656 | 75 | |
| GenBank mRNA | AB493697.1 | - | 4 - 5 | 427 | 75 | |
| AF005158.2 | - | 5 - 6 | 493 | 75 | ||
| BT025171.1 | - | 4 - 5 | 427 | 75 | ||
More Information
Set Membership: |
|

AGL24
Genomic Map