Assay Details
| Gene Symbol: AGL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118587.6 | NP_194185.1 | 8 - 9 | 879 | 71 | |
| GenBank mRNA | AF005158.2 | - | 8 - 9 | 716 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL24| Gene Symbol: AGL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118587.6 | NP_194185.1 | 8 - 9 | 879 | 71 | |
| GenBank mRNA | AF005158.2 | - | 8 - 9 | 716 | 71 | |
Set Membership: |
|