Assay Details
| Gene Symbol: PUM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036643.2 | NP_001031720.1 | 6 - 7 | 2121 | 67 | |
| NM_001341767.1 | NP_001329827.1 | 5 - 6 | 2079 | 67 | ||
| NM_001341768.1 | NP_001329828.1 | 5 - 6 | 2079 | 67 | ||
| NM_118721.3 | NP_567733.1 | 6 - 7 | 2400 | 67 | ||
| NM_202889.3 | NP_974618.2 | 6 - 7 | 2201 | 67 | ||
| GenBank mRNA | AF360298.1 | - | 6 - 7 | 2216 | 67 | |
| AK319148.1 | - | 5 - 6 | 2570 | 67 | ||
| AY133866.1 | - | 5 - 6 | 2079 | 67 | ||
More Information
Set Membership: |
|

PUM6
Genomic Map