Assay Details
| Gene Symbol: PUM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341768.1 | NP_001329828.1 | 8 - 9 | 2557 | 115 | |
| NM_118721.3 | NP_567733.1 | 9 - 10 | 2878 | 115 | ||
| NM_202889.3 | NP_974618.2 | 9 - 10 | 2679 | 115 | ||
| GenBank mRNA | AF360298.1 | - | 9 - 10 | 2694 | 115 | |
| AK319148.1 | - | 8 - 9 | 3048 | 115 | ||
| AY133866.1 | - | 8 - 9 | 2557 | 115 | ||
More Information
Set Membership: |
|

PUM6
Genomic Map