Assay Details
| Gene Symbol: CBL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118790.2 | NP_194386.1 | 1 - 2 | 233 | 69 | |
| GenBank mRNA | AF290434.1 | - | 1 - 2 | 119 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CBL7| Gene Symbol: CBL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118790.2 | NP_194386.1 | 1 - 2 | 233 | 69 | |
| GenBank mRNA | AF290434.1 | - | 1 - 2 | 119 | 69 | |
Set Membership: |
|