Assay Details
| Gene Symbol: CBL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341813.1 | NP_001320073.1 | 3 - 4 | 662 | 136 | |
| NM_118791.4 | NP_194387.1 | 3 - 4 | 450 | 136 | ||
| GenBank mRNA | AF076253.1 | - | 3 - 4 | 328 | 136 | |
| AY072441.1 | - | 3 - 4 | 343 | 136 | ||
| AY085344.1 | - | 3 - 4 | 331 | 136 | ||
| AY128880.1 | - | 1 - 2 | 127 | 136 | ||
More Information
Set Membership: |
|

CBL3
Genomic Map