Assay Details
| Gene Symbol: CBL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341813.1 | NP_001320073.1 | 8 - 9 | 1166 | 79 | |
| NM_118791.4 | NP_194387.1 | 8 - 9 | 942 | 79 | ||
| GenBank mRNA | AF076253.1 | - | 8 - 9 | 820 | 79 | |
| AY072441.1 | - | 8 - 9 | 835 | 79 | ||
| AY085344.1 | - | 8 - 9 | 835 | 79 | ||
| AY128880.1 | - | 6 - 7 | 619 | 79 | ||
More Information
Set Membership: |
|

CBL3
Genomic Map