Assay Details
| Gene Symbol: PDIL5-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118842.4 | NP_567765.2 | 11 - 12 | 1125 | 62 | |
| NM_148376.2 | NP_680742.2 | 12 - 13 | 1157 | 62 | ||
| GenBank mRNA | AY054496.1 | - | 11 - 12 | 1000 | 62 | |
| AY093266.1 | - | 11 - 12 | 917 | 62 | ||
More Information
Set Membership: |
|

PDIL5-4
Genomic Map