Assay Details
| Gene Symbol: PDIL5-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118842.4 | NP_567765.2 | 2 - 3 | 340 | 79 | |
| NM_148376.2 | NP_680742.2 | 3 - 4 | 372 | 79 | ||
| GenBank mRNA | AY054496.1 | - | 2 - 3 | 215 | 79 | |
| AY093266.1 | - | 2 - 3 | 132 | 79 | ||
More Information
Set Membership: |
|

PDIL5-4
Genomic Map