Assay Details
| Gene Symbol: NARA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118896.3 | NP_567780.1 | 1 - 2 | 519 | 131 | |
| GenBank mRNA | AY052355.1 | - | 1 - 2 | 497 | 131 | |
| AY059658.1 | - | 1 - 2 | 390 | 131 | ||
More Information
Set Membership: |
|

NARA5
Genomic Map