Assay Details
| Gene Symbol: AT3G54470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148875.5 | NP_680130.1 | 3 - 4 | 1033 | 92 | |
| GenBank mRNA | AY140098.1 | - | 3 - 4 | 1010 | 92 | |
| BT006612.1 | - | 3 - 4 | 958 | 92 | ||
More Information
Set Membership: |
|

AT3G54470
Genomic Map