Assay Details
| Gene Symbol: SYP121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112015.3 | NP_187788.1 | 1 - 2 | 957 | 91 | |
| NM_202559.3 | NP_974288.1 | 2 - 3 | 840 | 91 | ||
| GenBank mRNA | AF112864.1 | - | 1 - 2 | 638 | 91 | |
| AY087842.1 | - | 1 - 2 | 822 | 91 | ||
| AY093151.1 | - | 1 - 2 | 662 | 91 | ||
| BT003393.1 | - | 1 - 2 | 574 | 91 | ||
More Information
Set Membership: |
|

SYP121
Genomic Map