Assay Details
| Gene Symbol: NF-YC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112083.4 | NP_187854.2 | 4 - 5 | 640 | 72 | |
| GenBank mRNA | AF462841.1 | - | 4 - 5 | 566 | 72 | |
| AY143847.1 | - | 4 - 5 | 338 | 72 | ||
More Information
Set Membership: |
|

NF-YC11
Genomic Map