Assay Details
| Gene Symbol: TRFL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338001.1 | NP_001327482.1 | 2 - 3 | 476 | 119 | |
| NM_112091.4 | NP_187862.1 | 2 - 3 | 439 | 119 | ||
| GenBank mRNA | AK227790.1 | - | 2 - 3 | 364 | 119 | |
| AY550302.1 | - | 1 - 2 | 77 | 119 | ||
| BT006452.1 | - | 1 - 2 | 77 | 119 | ||
More Information
Set Membership: |
|

TRFL9
Genomic Map