Assay Details
| Gene Symbol: TRFL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338001.1 | NP_001327482.1 | 5 - 6 | 1452 | 138 | |
| NM_112091.4 | NP_187862.1 | 5 - 6 | 1415 | 138 | ||
| GenBank mRNA | AK227790.1 | - | 5 - 6 | 1340 | 138 | |
| AY550302.1 | - | 4 - 5 | 1053 | 138 | ||
| BT006452.1 | - | 4 - 5 | 1053 | 138 | ||
More Information
Set Membership: |
|

TRFL9
Genomic Map