Assay Details
| Gene Symbol: EMB2757 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119132.3 | NP_194715.2 | 8 - 9 | 814 | 89 | |
| GenBank mRNA | AB191306.1 | - | 8 - 9 | 790 | 89 | |
More Information
Set Membership: |
|

EMB2757
Genomic Map