Assay Details
| Gene Symbol: GAE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119190.4 | NP_194773.1 | 1 | 1543 | 104 | |
| GenBank mRNA | AY056303.1 | - | 1 | 1380 | 104 | |
| AY085505.1 | - | 1 | 1383 | 104 | ||
| AY099855.1 | - | 1 | 1351 | 104 | ||
| BT000308.1 | - | 1 | 1225 | 104 | ||
More Information
Set Membership: |
|

GAE1
Genomic Map