Assay Details
| Gene Symbol: TPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036679.1 | NP_001031756.1 | 1 - 2 | 448 | 82 | |
| NM_119194.5 | NP_194777.3 | 1 - 2 | 441 | 82 | ||
| NM_179226.3 | NP_849557.1 | 1 - 2 | 441 | 82 | ||
| GenBank mRNA | AF386955.1 | - | 1 - 2 | 380 | 82 | |
| AK316721.1 | - | 1 - 2 | 383 | 82 | ||
| BT006252.1 | - | 1 - 2 | 294 | 82 | ||
More Information
Set Membership: |
|

TPR1
Genomic Map