Assay Details
| Gene Symbol: SVL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124930.4 | NP_200359.1 | 2 - 3 | 564 | 90 | |
| GenBank mRNA | BT015819.1 | - | 2 - 3 | 476 | 90 | |
| BT020207.1 | - | 2 - 3 | 429 | 90 | ||
More Information
Set Membership: |
|

SVL1
Genomic Map