Assay Details
| Gene Symbol: CBL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341813.1 | NP_001320073.1 | 5 - 6 | 800 | 154 | |
| GenBank mRNA | AY085344.1 | - | 5 - 6 | 469 | 154 | |
More Information
Set Membership: |
|

CBL3
Genomic Map