Assay Details
| Gene Symbol: THM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179254.7 | NP_849585.1 | 1 - 2 | 425 | 63 | |
| GenBank mRNA | AF095749.1 | - | 1 - 2 | 264 | 63 | |
| BT004295.1 | - | 1 - 2 | 281 | 63 | ||
| BT005525.1 | - | 1 - 2 | 226 | 63 | ||
More Information
Set Membership: |
|

THM1
Genomic Map