Assay Details
| Gene Symbol: SERAT2;2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112150.4 | NP_187918.1 | 1 | 1644 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SERAT2;2| Gene Symbol: SERAT2;2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112150.4 | NP_187918.1 | 1 | 1644 | 116 | |
Set Membership: |
|