Assay Details
| Gene Symbol: ENP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342117.1 | NP_001327983.1 | 2 - 3 | 1731 | 62 | |
| NM_119332.4 | NP_194910.2 | 2 - 3 | 1135 | 62 | ||
| GenBank mRNA | AY150514.1 | - | 2 - 3 | 136 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001342117.1 transcripts.
Set Membership: |
|

ENP
Genomic Map