Assay Details
| Gene Symbol: GPX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342119.1 | NP_001328811.1 | 1 - 2 | 367 | 108 | |
| NM_119337.3 | NP_194915.2 | 1 - 2 | 367 | 108 | ||
More Information
Set Membership: |
|

GPX7
Genomic Map