Assay Details
| Gene Symbol: SHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119392.4 | NP_194966.1 | 1 - 2 | 879 | 82 | |
| GenBank mRNA | AF372944.1 | - | 1 - 2 | 589 | 82 | |
| AY074831.1 | - | 1 - 2 | 546 | 82 | ||
| AY084938.1 | - | 1 - 2 | 604 | 82 | ||
More Information
Set Membership: |
|

SHS1
Genomic Map