Assay Details
| Gene Symbol: SQS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342297.1 | NP_001329728.1 | 11 - 12 | 1182 | 108 | |
| NM_119630.4 | NP_195190.1 | 11 - 12 | 1428 | 108 | ||
| GenBank mRNA | AY099868.1 | - | 11 - 12 | 1231 | 108 | |
| BT003419.1 | - | 11 - 12 | 1128 | 108 | ||
| D29017.1 | - | 11 - 12 | 1342 | 108 | ||
| U79159.1 | - | 11 - 12 | 1255 | 108 | ||
| X86692.1 | - | 1241 | 108 | |||
More Information
Set Membership: |
|

SQS1
Genomic Map