Assay Details
| Gene Symbol: SQS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342297.1 | NP_001329728.1 | 2 - 3 | 293 | 70 | |
| NM_119630.4 | NP_195190.1 | 2 - 3 | 539 | 70 | ||
| GenBank mRNA | AY099868.1 | - | 2 - 3 | 342 | 70 | |
| BT003419.1 | - | 2 - 3 | 239 | 70 | ||
| D29017.1 | - | 2 - 3 | 453 | 70 | ||
| U79159.1 | - | 2 - 3 | 366 | 70 | ||
| X86692.1 | - | 352 | 70 | |||
More Information
Set Membership: |
|

SQS1
Genomic Map