Assay Details
| Gene Symbol: RAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203987.1 | NP_001190916.1 | 7 - 8 | 945 | 60 | |
| NM_001203988.2 | NP_001190917.1 | 7 - 8 | 713 | 60 | ||
| NM_001342313.1 | NP_001327974.1 | 6 - 7 | 811 | 60 | ||
| NM_001342314.1 | NP_001327973.1 | 4 - 5 | 762 | 60 | ||
| NM_119668.6 | NP_195228.1 | 5 - 6 | 685 | 60 | ||
| GenBank mRNA | AF031427.2 | - | 5 - 6 | 595 | 60 | |
| BT020318.1 | - | 5 - 6 | 487 | 60 | ||
| BT020603.1 | - | 5 - 6 | 428 | 60 | ||
More Information
Set Membership: |
|

RAC3
Genomic Map