Assay Details
| Gene Symbol: TPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334989.1 | NP_001319429.1 | 6 - 7 | 1119 | 64 | |
| NM_179582.2 | NP_849913.2 | 6 - 7 | 1064 | 64 | ||
More Information
Set Membership: |
|

TPR1
Genomic Map