Assay Details
| Gene Symbol: PIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335061.1 | NP_001323732.1 | 1 - 2 | 1629 | 76 | |
| NM_179592.2 | NP_849923.1 | 1 - 2 | 1754 | 76 | ||
| GenBank mRNA | AY091009.1 | - | 1 - 2 | 1313 | 76 | |
| BT008691.1 | - | 1 - 2 | 1318 | 76 | ||
More Information
Set Membership: |
|

PIN4
Genomic Map