Assay Details
| Gene Symbol: AGO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179779.3 | NP_850110.1 | 13 - 14 | 2179 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGO5| Gene Symbol: AGO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179779.3 | NP_850110.1 | 13 - 14 | 2179 | 150 | |
Set Membership: |
|