Assay Details
| Gene Symbol: VHA-A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179791.2 | NP_850122.1 | 16 - 17 | 2431 | 92 | |
| GenBank mRNA | AY091008.1 | - | 16 - 17 | 2335 | 92 | |
More Information
Set Membership: |
|

VHA-A1
Genomic Map