Assay Details
| Gene Symbol: AT3G15920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112462.6 | NP_188213.1 | 1 - 2 | 3535 | 73 | |
| GenBank mRNA | AK175543.1 | - | 1 - 2 | 319 | 73 | |
| AK176000.1 | - | 1 - 2 | 327 | 73 | ||
| BT012581.1 | - | 1 - 2 | 271 | 73 | ||
More Information
Set Membership: |
|

AT3G15920
Genomic Map