Assay Details
| Gene Symbol: AT3G15920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112462.6 | NP_188213.1 | 2 - 3 | 3647 | 66 | |
| GenBank mRNA | AK175543.1 | - | 2 - 3 | 431 | 66 | |
| AK176000.1 | - | 2 - 3 | 439 | 66 | ||
| BT012581.1 | - | 2 - 3 | 383 | 66 | ||
More Information
Set Membership: |
|

AT3G15920
Genomic Map