Assay Details
| Gene Symbol: AT3G15920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112462.6 | NP_188213.1 | 6 - 7 | 4645 | 75 | |
| GenBank mRNA | AK175543.1 | - | 6 - 7 | 1429 | 75 | |
| AK176000.1 | - | 6 - 7 | 1437 | 75 | ||
| BT012581.1 | - | 6 - 7 | 1381 | 75 | ||
More Information
Set Membership: |
|

AT3G15920
Genomic Map