Assay Details
| Gene Symbol: PUM24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112555.2 | NP_566559.1 | 10 - 11 | 1199 | 129 | |
| GenBank mRNA | AY045638.1 | - | 10 - 11 | 1156 | 129 | |
More Information
Set Membership: |
|

PUM24
Genomic Map