Assay Details
| Gene Symbol: PUM24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112555.2 | NP_566559.1 | 15 - 16 | 1672 | 80 | |
| GenBank mRNA | AY045638.1 | - | 15 - 16 | 1629 | 80 | |
More Information
Set Membership: |
|

PUM24
Genomic Map