Assay Details
| Gene Symbol: PUM24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112555.2 | NP_566559.1 | 3 - 4 | 451 | 82 | |
| GenBank mRNA | AY045638.1 | - | 3 - 4 | 408 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PUM24| Gene Symbol: PUM24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112555.2 | NP_566559.1 | 3 - 4 | 451 | 82 | |
| GenBank mRNA | AY045638.1 | - | 3 - 4 | 408 | 82 | |
Set Membership: |
|