Assay Details
| Gene Symbol: PUM24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112555.2 | NP_566559.1 | 7 - 8 | 858 | 59 | |
| GenBank mRNA | AY045638.1 | - | 7 - 8 | 815 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PUM24| Gene Symbol: PUM24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112555.2 | NP_566559.1 | 7 - 8 | 858 | 59 | |
| GenBank mRNA | AY045638.1 | - | 7 - 8 | 815 | 59 | |
Set Membership: |
|