Assay Details
| Gene Symbol: UBC32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112576.3 | NP_566563.1 | 2 - 3 | 610 | 109 | |
| GenBank mRNA | AF386935.1 | - | 2 - 3 | 360 | 109 | |
| BT020530.1 | - | 2 - 3 | 244 | 109 | ||
More Information
Set Membership: |
|

UBC32
Genomic Map