Assay Details
| Gene Symbol: UBC32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112576.3 | NP_566563.1 | 4 - 5 | 862 | 70 | |
| GenBank mRNA | AF386935.1 | - | 4 - 5 | 612 | 70 | |
| BT020530.1 | - | 4 - 5 | 496 | 70 | ||
More Information
Set Membership: |
|

UBC32
Genomic Map