Assay Details
| Gene Symbol: UPL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125176.1 | NP_001118648.1 | 22 - 23 | 2982 | 134 | |
| NM_001202986.1 | NP_001189915.1 | 22 - 23 | 3065 | 134 | ||
| NM_112597.4 | NP_188346.2 | 22 - 23 | 3267 | 134 | ||
| GenBank mRNA | AY093202.1 | - | 22 - 23 | 3025 | 134 | |
| BT010593.1 | - | 22 - 23 | 2790 | 134 | ||
More Information
Set Membership: |
|

UPL6
Genomic Map