Assay Details
| Gene Symbol: CRSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338291.1 | NP_001327079.1 | 1 - 2 | 432 | 87 | |
| NM_001338292.1 | NP_001327078.1 | 1 - 2 | 421 | 87 | ||
| NM_112627.4 | NP_188374.2 | 1 - 2 | 412 | 87 | ||
| GenBank mRNA | AK228656.1 | - | 1 - 2 | 412 | 87 | |
| BT006180.1 | - | 1 - 2 | 412 | 87 | ||
More Information
Set Membership: |
|

CRSH
Genomic Map