Assay Details
| Gene Symbol: CIPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112631.3 | NP_566580.1 | 5 - 6 | 892 | 133 | |
| NM_202599.2 | NP_974328.1 | 3 - 4 | 1141 | 133 | ||
| GenBank mRNA | BT029495.1 | - | 3 - 4 | 278 | 133 | |
More Information
Set Membership: |
|

CIPK1
Genomic Map