Assay Details
| Gene Symbol: CIPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112631.3 | NP_566580.1 | 6 - 7 | 1064 | 64 | |
| NM_202599.2 | NP_974328.1 | 4 - 5 | 1313 | 64 | ||
| GenBank mRNA | BT029495.1 | - | 4 - 5 | 450 | 64 | |
More Information
Set Membership: |
|

CIPK1
Genomic Map